missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Fluorophenyl)valeric acid, 97%

CAS: 24484-22-8 | C11H12FO2 | 195.21 g/mol

Supplier:  Thermo Scientific Chemicals L1793506

Quantity 5 g

 View more versions of this product

Catalog No. AAL1793506


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

24484-22-8
195.21
BXEFPLJKYWEEAN-UHFFFAOYSA-M
5-(4-fluorophenyl)pentanoic acid
C11H12FO2
MFCD00800242
2734670
[O-]C(=O)CCCCC1=CC=C(F)C=C1

Specifications

5-(4-Fluorophenyl)valeric acid
73°C to 75°C
5 g
7918899
BXEFPLJKYWEEAN-UHFFFAOYSA-M
5-(4-fluorophenyl)pentanoic acid
2734670
97%
24484-22-8
C11H12FO2
MFCD00800242
5-4-fluorophenyl valeric acid, 5-4-fluorophenyl pentanoic acid, benzenepentanoic acid, 4-fluoro, 5-4'-fluorophenyl valeric acid, acmc-1cj6r, benzenepentanoic acid,4-fluoro, 5-4-fluorophenyl-pentanoic acid, 5-4-fluoro-phenyl-pentanoic acid
[O-]C(=O)CCCCC1=CC=C(F)C=C1
195.21
196.22
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions