Learn More
5-(4-Fluorophenyl)valeric acid, 97%
CAS: 24484-22-8 | C11H12FO2 | 195.21 g/mol
Supplier: Thermo Scientific Chemicals L1793503
| Quantity | 1 g |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 24484-22-8 | |
| 195.21 | |
| BXEFPLJKYWEEAN-UHFFFAOYSA-M | |
| 5-(4-fluorophenyl)pentanoic acid |
| C11H12FO2 | |
| MFCD00800242 | |
| 2734670 | |
| [O-]C(=O)CCCCC1=CC=C(F)C=C1 |
Specifications
| 5-(4-Fluorophenyl)valeric acid | |
| 73°C to 75°C | |
| 1 g | |
| 7918899 | |
| BXEFPLJKYWEEAN-UHFFFAOYSA-M | |
| 5-(4-fluorophenyl)pentanoic acid | |
| 2734670 | |
| 97% |
| 24484-22-8 | |
| C11H12FO2 | |
| MFCD00800242 | |
| 5-4-fluorophenyl valeric acid, 5-4-fluorophenyl pentanoic acid, benzenepentanoic acid, 4-fluoro, 5-4'-fluorophenyl valeric acid, acmc-1cj6r, benzenepentanoic acid,4-fluoro, 5-4-fluorophenyl-pentanoic acid, 5-4-fluoro-phenyl-pentanoic acid | |
| [O-]C(=O)CCCCC1=CC=C(F)C=C1 | |
| 195.21 | |
| 196.22 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures