missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Fluorophenyl)thiophene-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. F10731G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10731G 1 g
F10735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. F10731G missing translation for 'mfr' TCI America missing translation for 'supplierNo' F10731G

Chemical Identifiers

CAS 115933-30-7
Molecular Formula C11H7FO2S
Molecular Weight (g/mol) 222.233
MDL Number MFCD01316570
InChI Key YOFXYDVYMHOXSY-UHFFFAOYSA-N
Synonym 5-4-fluorophenyl thiophene-2-carboxylic acid, 5-4-fluorophenyl-2-thiophenecarboxylic acid, 2-carboxy-5-4-fluorophenyl thiophene, 2-thiophenecarboxylicacid, 5-4-fluorophenyl, 5-4-fluoro-phenyl-thiophene-2-carboxylicacid, fluorophenylthiophenecarboxylicacid, 5-4-fluorophenyl-2-thenoic acid, 1-5-carboxythien-2-yl-4-fluorobenzene, 5-4-fluorophenyl thiophen-2-carboxylic acid, 5-4-fluorophenyl thiophene-2-carboxylicacid
PubChem CID 1873401
IUPAC Name 5-(4-fluorophenyl)thiophene-2-carboxylic acid
SMILES C1=CC(=CC=C1C2=CC=C(S2)C(=O)O)F

Specifications

Melting Point 250°C
Color White-Yellow
Quantity 1 g
Formula Weight 222.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(4-Fluorophenyl)thiophene-2-carboxylic Acid