Learn More
Thermo Scientific Chemicals 5-(4-Dimethylaminobenzylidene)rhodanine, 99%
5-(4-Dimethylaminobenzylidene)rhodanine, 99%, Quantity: 100g, Packaging: Glass bottle, Red, Melting Point: 285 deg.C to 288 deg.C, Molecular Weight: 264.35, Percent Purity: 99%, Appearance: (0.3% soln. in acetone) clear, Assay Percent Range: 99%, Beilstein: 27, 433, CAS: 536-17-4 | CAS: 536-17-4 | C12H12N2OS2 | 264.35 g/mol
Supplier: Thermo Scientific Chemicals 151470250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 5-(4-Dimethylaminobenzylidene)rhodanine | |
| 98.5 | |
| 285°C to 288°C | |
| Authentic | |
| Glass bottle | |
| MFCD00064857 | |
| 27, 433 | |
| unii-0ser53q7rt, p-dimethylaminobenzalrhodanine, 5-4-dimethylaminobenzylidene rhodanine, 0ser53q7rt, usaf pd-20, 4-dimethylaminobenzylidenerhodanine, 4-thiazolidinone, 5-4-dimethylamino phenyl methylene-2-thioxo, 5-p-dimethylaminobenzal rhodanine, p-dimethylaminobenzylidene rhodamine, p-dimethylamino benzal-5-rhodanine | |
| JJRVRELEASDUMY-UHFFFAOYSA-N | |
| 5-[[4-(dimethylamino)phenyl]methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one | |
| 2723826 | |
| 99% |
| 536-17-4 | |
| 100.0 | |
| Red | |
| 99% | |
| C12H12N2OS2 | |
| 25 g | |
| 15, 3253 | |
| (0.03% in acetone) Clear | |
| CN(C)C1=CC=C(C=C1)C=C2C(=O)NC(=S)S2 | |
| 264.35 | |
| 264.35 | |
| Fluffy Powder |
Chemical Identifiers
| 536-17-4 | |
| 264.35 | |
| JJRVRELEASDUMY-UHFFFAOYSA-N | |
| 2723826 | |
| CN(C)C1=CC=C(C=C1)C=C2C(=O)NC(=S)S2 |
| C12H12N2OS2 | |
| MFCD00064857 | |
| unii-0ser53q7rt, p-dimethylaminobenzalrhodanine, 5-4-dimethylaminobenzylidene rhodanine, 0ser53q7rt, usaf pd-20, 4-dimethylaminobenzylidenerhodanine, 4-thiazolidinone, 5-4-dimethylamino phenyl methylene-2-thioxo, 5-p-dimethylaminobenzal rhodanine, p-dimethylaminobenzylidene rhodamine, p-dimethylamino benzal-5-rhodanine | |
| 5-[[4-(dimethylamino)phenyl]methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one |
Safety and Handling
EINECSNumber : 208-625-2
RTECSNumber : VI8090000
TSCA : TSCA
RUO – Research Use Only