missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 5-(4-Dimethylaminobenzylidene)rhodanine, 99%

Catalog No. AC151470100
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC151470100 10 g Glass bottle
AC151470250 25 g Glass bottle
AC151471000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC151470100 Supplier Thermo Scientific Chemicals Supplier No. 151470100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

5-(4-Dimethylaminobenzylidene)rhodanine, 99%, Quantity: 100g, Packaging: Glass bottle, Red, Melting Point: 285 deg.C to 288 deg.C, Molecular Weight: 264.35, Percent Purity: 99%, Appearance: (0.3% soln. in acetone) clear, Assay Percent Range: 99%, Beilstein: 27, 433, CAS: 536-17-4 | CAS: 536-17-4 | C12H12N2OS2 | 264.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 536-17-4
Molecular Formula C12H12N2OS2
Molecular Weight (g/mol) 264.35
MDL Number MFCD00064857
InChI Key JJRVRELEASDUMY-UHFFFAOYSA-N
Synonym unii-0ser53q7rt, p-dimethylaminobenzalrhodanine, 5-4-dimethylaminobenzylidene rhodanine, 0ser53q7rt, usaf pd-20, 4-dimethylaminobenzylidenerhodanine, 4-thiazolidinone, 5-4-dimethylamino phenyl methylene-2-thioxo, 5-p-dimethylaminobenzal rhodanine, p-dimethylaminobenzylidene rhodamine, p-dimethylamino benzal-5-rhodanine
PubChem CID 2723826
IUPAC Name 5-[[4-(dimethylamino)phenyl]methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
SMILES CN(C)C1=CC=C(C=C1)C=C2C(=O)NC(=S)S2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 285°C to 288°C
Color Red
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 10 g
Beilstein 27, 433
Merck Index 15, 3253
Solubility Information (0.03% in acetone) Clear
Formula Weight 264.35
Percent Purity 99%
Physical Form Fluffy Powder
Chemical Name or Material 5-(4-Dimethylaminobenzylidene)rhodanine
Show More Show Less
EINECSNumber 208-625-2
RTECSNumber VI8090000
TSCA TSCA

RUO – Research Use Only