missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Dimethylaminobenzylidene)rhodanine 98%

Catalog No. AAA1715206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1715206 5 g
AAA1715214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1715206 Supplier Thermo Scientific Chemicals Supplier No. A1715206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 536-17-4 | C12H12N2OS2 | 264.361 g/mol

Applied for the detection of Ag, Au, Cu, Hg, Pd, Pt, alkaloids, antipyrin, indican, sulfonamide, urobilinogen. Reagent for the determination of silver, palladium, mercury and gold. Also used in the photometric determination of auto radiograms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Applied for the detection of Ag, Au, Cu, Hg, Pd, Pt, alkaloids, antipyrin, indican, sulfonamide, urobilinogen. Reagent for the determination of silver, palladium, mercury and gold. Also used in the photometric determination of auto radiograms.

Solubility
Insoluble in water, 993.7(mg/L) at 25°C

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.

Chemical Identifiers

CAS 536-17-4
Molecular Formula C12H12N2OS2
Molecular Weight (g/mol) 264.361
MDL Number MFCD00064857
InChI Key JJRVRELEASDUMY-UHFFFAOYSA-N
Synonym unii-0ser53q7rt, p-dimethylaminobenzalrhodanine, 5-4-dimethylaminobenzylidene rhodanine, 0ser53q7rt, usaf pd-20, 4-dimethylaminobenzylidenerhodanine, 4-thiazolidinone, 5-4-dimethylamino phenyl methylene-2-thioxo, 5-p-dimethylaminobenzal rhodanine, p-dimethylaminobenzylidene rhodamine, p-dimethylamino benzal-5-rhodanine
PubChem CID 2723826
IUPAC Name 5-[[4-(dimethylamino)phenyl]methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
SMILES CN(C)C1=CC=C(C=C1)C=C2C(=O)NC(=S)S2

Specifications

Melting Point 275°C to 280°C
Odor Weak
Quantity 5 g
Beilstein 189065
Merck Index 14,3231
Solubility Information Insoluble in water,993.7(mg/L) at 25°C
Formula Weight 264.37
Percent Purity 98%
Chemical Name or Material 5-(4-Dimethylaminobenzylidene)rhodanine 98%
EINECSNumber 208-625-2
RTECSNumber VI8090000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only