missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Carboxyphenyl)-10,15,20-triphenylporphyrin 98.0+%, TCI America™

Catalog No. C1630100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1630100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C1630100MG Supplier TCI America Supplier No. C1630100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95051-10-8
Molecular Formula C45H30N4O2
Molecular Weight (g/mol) 658.76
MDL Number MFCD01366156
InChI Key VPIIZALKBPNPDZ-UHFFFAOYSA-N
Synonym 5-(4-Carboxyphenyl)-10,15,20-triphenyl-21H,23H-porphine
PubChem CID 6321175
IUPAC Name 4-{7,12,17-triphenyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3(24),4,6,8,10,12,14,16,18(21),19-undecaen-2-yl}benzoic acid
SMILES OC(=O)C1=CC=C(C=C1)C1=C2C=CC(=N2)C(=C2NC(C=C2)=C(C2=CC=C(N2)C(=C2C=CC1=N2)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Purple
Quantity 100 mg
Formula Weight 658.76
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-(4-Carboxyphenyl)-10,15,20-triphenylporphyrin