missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Bromophenyl)isoxazole, 97%, Thermo Scientific™

Catalog No. CC12910DA
Change view
Click to view available options
Quantity:
1g
10g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC12910DA 1g Amber Glass Bottle
CC12910EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC12910DA Supplier Thermo Scientific Chemicals Supplier No. CC12910DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7064-31-5
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.057
MDL Number MFCD02183532
InChI Key AVMMSHZGFRWLRI-UHFFFAOYSA-N
Synonym 5-4-bromophenyl isoxazole, 5-4-bromophenyl-1,2-oxazole, acmc-209ofh
PubChem CID 2735608
IUPAC Name 5-(4-bromophenyl)-1,2-oxazole
SMILES C1=CC(=CC=C1C2=CC=NO2)Br

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 224.06
Percent Purity 95%
Chemical Name or Material 5-(4-Bromophenyl)isoxazole