missing translation for 'onlineSavingsMsg'
Learn More

5(4)-Amino-4(5)-(aminocarbonyl)imidazole Hydrochloride 98.0+%, TCI America™

Catalog No. A11755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11755G 5 g
A117525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11755G Supplier TCI America Supplier No. A11755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 72-40-2
Molecular Formula C4H7ClN4O
Molecular Weight (g/mol) 162.58
MDL Number MFCD00012704
InChI Key MXCUYSMIELHIQL-UHFFFAOYSA-N
Synonym 4-amino-5-imidazolecarboxamide hydrochloride, 5-amino-4-imidazolecarboxamide hydrochloride, 5-amino-1h-imidazole-4-carboxamide hydrochloride, 5-aminoimidazole-4-carboxamide hydrochloride, aic .cntdot. hcl, 4-amino-1h-imidazole-5-carboxamide hydrochloride, aminoimidazolecarboxamide hydrochloride, 4-aminoimidazole-5-carboxamide hydrochloride, 1h-imidazole-4-carboxamide, 5-amino-, monohydrochloride, 5-aminoimidazol-4-carboxamide, hydrochloride
PubChem CID 66146
IUPAC Name 4-amino-1H-imidazole-5-carboxamide hydrochloride
SMILES Cl.NC(=O)C1=C(N)N=CN1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 162.58
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5(4)-Amino-4(5)-(aminocarbonyl)imidazole Hydrochloride
RTECSNumber NI3911000
TSCA Yes