missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,2'-bithiophene 98.0+%, TCI America™
Supplier: TCI America T251825G
Specifications
| 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,2′-bithiophene | |
| Blue-Green | |
| MFCD04039974 | |
| 2,2'-bithiophene-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,2'-bithiophene, 2-2,2'-bithiophen-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-5-thiophen-2-yl thiophen-2-yl-1,3,2-dioxaborolane, acmc-209kb7, 4,4,5,5-tetramethyl-2-5-thiophen-2-ylthiophen-2-yl-1,3,2-dioxaborolane, 2,2'-bithiophene-5'-boronic acid pinacol ester | |
| CC1(C)OB(OC1(C)C)C1=CC=C(S1)C1=CC=CS1 | |
| 292.22 | |
| 292.22 | |
| Crystalline Lumps |
| 479719-88-5 | |
| C14H17BO2S2 | |
| 25 g | |
| HPOQARMSOPOZMW-UHFFFAOYSA-N | |
| 2-{[2,2'-bithiophen]-5-yl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 3592790 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 479719-88-5 | |
| 292.22 | |
| HPOQARMSOPOZMW-UHFFFAOYSA-N | |
| 3592790 | |
| CC1(C)OB(OC1(C)C)C1=CC=C(S1)C1=CC=CS1 |
| C14H17BO2S2 | |
| MFCD04039974 | |
| 2,2'-bithiophene-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,2'-bithiophene, 2-2,2'-bithiophen-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-5-thiophen-2-yl thiophen-2-yl-1,3,2-dioxaborolane, acmc-209kb7, 4,4,5,5-tetramethyl-2-5-thiophen-2-ylthiophen-2-yl-1,3,2-dioxaborolane, 2,2'-bithiophene-5'-boronic acid pinacol ester | |
| 2-{[2,2'-bithiophen]-5-yl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No