missing translation for 'onlineSavingsMsg'
Learn More

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,2'-bithiophene 98.0+%, TCI America™

Catalog No. T25181G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25181G 1 g
T25185G 5 g
T251825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T25181G Supplier TCI America Supplier No. T25181G

Chemical Identifiers

CAS 479719-88-5
Molecular Formula C14H17BO2S2
Molecular Weight (g/mol) 292.22
MDL Number MFCD04039974
InChI Key HPOQARMSOPOZMW-UHFFFAOYSA-N
Synonym 2,2'-bithiophene-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,2'-bithiophene, 2-2,2'-bithiophen-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-5-thiophen-2-yl thiophen-2-yl-1,3,2-dioxaborolane, acmc-209kb7, 4,4,5,5-tetramethyl-2-5-thiophen-2-ylthiophen-2-yl-1,3,2-dioxaborolane, 2,2'-bithiophene-5'-boronic acid pinacol ester
PubChem CID 3592790
IUPAC Name 2-{[2,2'-bithiophen]-5-yl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(S1)C1=CC=CS1

Specifications

Color Blue-Green
Quantity 1 g
Formula Weight 292.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,2′-bithiophene