missing translation for 'onlineSavingsMsg'
Learn More

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole 98.0+%, TCI America™

Catalog No. I05901G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05901G 1 g
I05905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05901G Supplier TCI America Supplier No. I05901G
Only null left

Chemical Identifiers

CAS 269410-24-4
Molecular Formula C14H18BNO2
Molecular Weight (g/mol) 243.11
MDL Number MFCD03789263
InChI Key FATPQDPUKVVCLT-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 5-indoleboronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, indole-5-boronic acid pinacol ester, 1h-indole-5-boronic acid pinacol ester, 5-indolylboronic acid, pinacol ester, 1h-indole, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indole, indolyl-5-boronic acid pinacol ester, indole-5-boronic acid, pinacol ester
PubChem CID 2734772
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
SMILES CC1(C)OB(OC1(C)C)C1=CC=C2NC=CC2=C1

Specifications

Melting Point 126°C
Quantity 1 g
Formula Weight 243.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole