missing translation for 'onlineSavingsMsg'
Learn More

5-(3-Pyridinyloxy)-2-furoic acid, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals AW01246DE

 View more versions of this product

Catalog No. AW01246DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

5-(3-Pyridinyloxy)-2-furoic acid
96.5
97%
MFCD03407385
5-3-pyridinyloxy-2-furoic acid, 5-pyridin-3-yloxy furan-2-carboxylic acid, 2-furancarboxylic acid,5-3-pyridinyloxy, 5-3-pyridyloxy furan-2-carboxylic acid, 5-3-pyridinyl-oxy-2-furoic acid, 5-pyridin-3-yl oxy furan-2-carboxylic acid
C1=CC(=CN=C1)OC2=CC=C(O2)C(=O)O
205.169
205.17
852180-39-3
100.0
C10H7NO4
5g
ZTYMSWYWSCYFAF-UHFFFAOYSA-N
5-pyridin-3-yloxyfuran-2-carboxylic acid
7060545
97%

Chemical Identifiers

852180-39-3
205.169
ZTYMSWYWSCYFAF-UHFFFAOYSA-N
7060545
C1=CC(=CN=C1)OC2=CC=C(O2)C(=O)O
C10H7NO4
MFCD03407385
5-3-pyridinyloxy-2-furoic acid, 5-pyridin-3-yloxy furan-2-carboxylic acid, 2-furancarboxylic acid,5-3-pyridinyloxy, 5-3-pyridyloxy furan-2-carboxylic acid, 5-3-pyridinyl-oxy-2-furoic acid, 5-pyridin-3-yl oxy furan-2-carboxylic acid
5-pyridin-3-yloxyfuran-2-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions