missing translation for 'onlineSavingsMsg'
Learn More

5-[3-(Bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole, 97%, Thermo Scientific™

Catalog No. p-3540994
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC49508CB 250mg Amber Glass Bottle
CC49508DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC49508CB Supplier Thermo Scientific Chemicals Supplier No. CC49508CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 926921-57-5
Molecular Formula C10H9BrN2O
Molecular Weight (g/mol) 253.099
MDL Number MFCD09817470
InChI Key WKTARCMEMVHRLR-UHFFFAOYSA-N
Synonym 5-3-bromomethyl phenyl-3-methyl-1,2,4-oxadiazole, 3-3-methyl-1,2,4-oxadiazol-5-yl benzyl bromide, 5-3-bromomethyl phenyl-3-methyl-1,2,4-oxadiazol, alpha-bromo-3-3-methyl-1,2,4-oxadiazol-5-yl toluene, 1-bromomethyl-3-3-methyl-1,2,4-oxadiazol-5-yl benzene, pubchem23332
PubChem CID 24229497
IUPAC Name 5-[3-(bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole
SMILES CC1=NOC(=N1)C2=CC(=CC=C2)CBr

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 253.1
Percent Purity 97%
Chemical Name or Material 5-[3-(Bromomethyl)phenyl]-3-methyl-1, 2, 4-oxadiazole
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.