missing translation for 'onlineSavingsMsg'
Learn More

5-[3-(Bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC49508DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC49508DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

926921-57-5
253.099
WKTARCMEMVHRLR-UHFFFAOYSA-N
24229497
CC1=NOC(=N1)C2=CC(=CC=C2)CBr
C10H9BrN2O
MFCD09817470
5-3-bromomethyl phenyl-3-methyl-1,2,4-oxadiazole, 3-3-methyl-1,2,4-oxadiazol-5-yl benzyl bromide, 5-3-bromomethyl phenyl-3-methyl-1,2,4-oxadiazol, alpha-bromo-3-3-methyl-1,2,4-oxadiazol-5-yl toluene, 1-bromomethyl-3-3-methyl-1,2,4-oxadiazol-5-yl benzene, pubchem23332
5-[3-(bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole

Specifications

5-[3-(Bromomethyl)phenyl]-3-methyl-1, 2, 4-oxadiazole
96.5
97%
C10H9BrN2O
MFCD09817470
WKTARCMEMVHRLR-UHFFFAOYSA-N
5-[3-(bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole
24229497
97%
926921-57-5
100.0
Amber Glass Bottle
1g
5-3-bromomethyl phenyl-3-methyl-1,2,4-oxadiazole, 3-3-methyl-1,2,4-oxadiazol-5-yl benzyl bromide, 5-3-bromomethyl phenyl-3-methyl-1,2,4-oxadiazol, alpha-bromo-3-3-methyl-1,2,4-oxadiazol-5-yl toluene, 1-bromomethyl-3-3-methyl-1,2,4-oxadiazol-5-yl benzene, pubchem23332
CC1=NOC(=N1)C2=CC(=CC=C2)CBr
253.099
253.1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents