missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Pyridyl)-1H-tetrazole, 98%

Catalog No. p-7025401
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAB2085203 1 g
AAB2085206 5 g
2 options

Catalog No. AAB2085203

Supplier: Thermo Scientific Chemicals B2085203

Only null left

CAS: 33893-89-9 | C6H4N5 | 146.13 g/mol

It is used in the synthesis and crystal structure of the [Ag(2-allyl-5-phenyl-2Htetrazole)ClO4] π,complex .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis and crystal structure of the [Ag(2-allyl-5-phenyl-2Htetrazole)ClO4] π,σ-complex .

Solubility
Soluble in water at 25°C 11 g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 33893-89-9
Molecular Formula C6H4N5
Molecular Weight (g/mol) 146.13
MDL Number MFCD00068114
InChI Key ZKHJXYLAESJIEI-UHFFFAOYSA-N
Synonym 5-2-pyridyl-1h-tetrazole, 2-1h-tetrazol-5-yl pyridine, 2-2h-tetrazol-5-yl pyridine, 2-2h-tetrazol-5-yl-pyridine, pyridine, 2-1h-tetrazol-5-yl, 5-2-pyridinyl tetrazole, 2-1h-1,2,3,4-tetrazol-5-yl pyridine, 5-2-pyridyl tetrazole, 5-pyridin-2-yl tetrazole, chembl80094
PubChem CID 320267
SMILES [N-]1N=NN=C1C1=CC=CC=N1

Specifications

Quantity 1 g
Solubility Information Soluble in water at 25°C 11g/L.
IUPAC Name 5-(pyridin-2-yl)-1H-1,2,3,4-tetrazol-1-ide
Formula Weight 147.14
Percent Purity 98%
Chemical Name or Material 5-(2-Pyridyl)-1H-tetrazole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only