missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Pyridyl)-1,3-oxazole, 97%

Catalog No. p-7032603
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34123MD 250 mg
AAH3412303 1 g
AAH3412306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH34123MD Supplier Thermo Scientific Chemicals Supplier No. H34123MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 70380-73-3 | C8H6N2O | 146.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 70380-73-3
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.149
MDL Number MFCD00085160
InChI Key LUMKXMDGONYZHO-UHFFFAOYSA-N
PubChem CID 737194
IUPAC Name 5-pyridin-2-yl-1,3-oxazole
SMILES C1=CC=NC(=C1)C2=CN=CO2

Specifications

Quantity 250 mg
Synonym 5-2-pyridyl-1,3-oxazole, 2-1,3-oxazol-5-yl pyridine, 5-pyridin-2-yl oxazole, pyridine,2-5-oxazolyl, 2-5-oxazolyl pyridine, 2-oxazol-5-yl-pyridine, 5-pyridin-2-yl-1,3-oxazole, pyridine, 2-5-oxazolyl, 5-2-pyridyl oxazole, acmc-209oeb
Formula Weight 146.15
Percent Purity 97%
Chemical Name or Material 5-(2-Pyridyl)-1,3-oxazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only