missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Methylphenyl)-1H-tetrazole, 99%

CAS: 51449-86-6 | C8H8N4 | 160.18 g/mol

$104.51 - $166.11

Chemical Identifiers

CAS 51449-86-6
Molecular Formula C8H8N4
Molecular Weight (g/mol) 160.18
MDL Number MFCD03093095
InChI Key MTBUOESFHRORQT-UHFFFAOYSA-N
Synonym 5-2-methylphenyl-1h-tetrazole, 5-o-tolyl-1h-tetrazole, 5-o-tolyl-2h-tetrazole, 5-2-methylphenyl-2h-tetrazole, 5-2-methylphenyl tetrazole, 5-2-methylphenyl-2h-1,2,3,4-tetrazole, 2h-tetrazole,5-2-methylphenyl, 5-2-methylphenyl-1h-1,2,3,4-tetrazole, 2-methylphenyltetrazole, 5-o-tolyl tetrazole
PubChem CID 583067
IUPAC Name 5-(2-methylphenyl)-2H-tetrazole
SMILES CC1=CC=CC=C1C2=NNN=N2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL19262MD
View Documents
Thermo Scientific Chemicals
L19262MD
250 mg
Each for $104.51
Only null left
 
AAL1926203
View Documents
Thermo Scientific Chemicals
L1926203
1 g
Each for $166.11
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

51449-86-6
160.18
MTBUOESFHRORQT-UHFFFAOYSA-N
583067
CC1=CC=CC=C1C2=NNN=N2
C8H8N4
MFCD03093095
5-2-methylphenyl-1h-tetrazole, 5-o-tolyl-1h-tetrazole, 5-o-tolyl-2h-tetrazole, 5-2-methylphenyl-2h-tetrazole, 5-2-methylphenyl tetrazole, 5-2-methylphenyl-2h-1,2,3,4-tetrazole, 2h-tetrazole,5-2-methylphenyl, 5-2-methylphenyl-1h-1,2,3,4-tetrazole, 2-methylphenyltetrazole, 5-o-tolyl tetrazole
5-(2-methylphenyl)-2H-tetrazole

Specifications

51449-86-6
C8H8N4
250 mg
5-2-methylphenyl-1h-tetrazole, 5-o-tolyl-1h-tetrazole, 5-o-tolyl-2h-tetrazole, 5-2-methylphenyl-2h-tetrazole, 5-2-methylphenyl tetrazole, 5-2-methylphenyl-2h-1,2,3,4-tetrazole, 2h-tetrazole,5-2-methylphenyl, 5-2-methylphenyl-1h-1,2,3,4-tetrazole, 2-methylphenyltetrazole, 5-o-tolyl tetrazole
CC1=CC=CC=C1C2=NNN=N2
160.18
160.18
5-(2-Methylphenyl)-1H-tetrazole
151°C to 154°C
MFCD03093095
136151
MTBUOESFHRORQT-UHFFFAOYSA-N
5-(2-methylphenyl)-2H-tetrazole
583067
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only