missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Methyl-1,3-thiazol-4-yl)-2-thiophenecarboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 400715-45-9
Molecular Formula C9H7NO2S2
Molecular Weight (g/mol) 225.28
MDL Number MFCD02682053
InChI Key AEDWMWQOFJYAGN-UHFFFAOYSA-N
Synonym 5-2-methyl-1,3-thiazol-4-yl thiophene-2-carboxylic acid, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenecarboxylic acid, 2-thiophenecarboxylicacid, 5-2-methyl-4-thiazolyl, 5-2-methylthiazol-4-yl thiophene-2-carboxylic acid, 5-2-methyl-1,3-thiazol-4-yl thiophen-2-carboxylic acid, 5-2-methylthiazol-4-yl-thiophene-2-carboxylic acid, 5-2-methylthiazol4-yl-thiophene-2-carboxylic acid, thiophene-2-carboxylic acid, 5-2-methyl-4-thiazolyl, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenecarboxylic acid #
PubChem CID 614467
IUPAC Name 5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-carboxylic acid
SMILES CC1=NC(=CS1)C2=CC=C(S2)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CC18201CB
View Documents
N/A 250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC18201DA
View Documents
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC18201DE
View Documents
N/A 5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

400715-45-9
225.28
AEDWMWQOFJYAGN-UHFFFAOYSA-N
614467
CC1=NC(=CS1)C2=CC=C(S2)C(=O)O
C9H7NO2S2
MFCD02682053
5-2-methyl-1,3-thiazol-4-yl thiophene-2-carboxylic acid, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenecarboxylic acid, 2-thiophenecarboxylicacid, 5-2-methyl-4-thiazolyl, 5-2-methylthiazol-4-yl thiophene-2-carboxylic acid, 5-2-methyl-1,3-thiazol-4-yl thiophen-2-carboxylic acid, 5-2-methylthiazol-4-yl-thiophene-2-carboxylic acid, 5-2-methylthiazol4-yl-thiophene-2-carboxylic acid, thiophene-2-carboxylic acid, 5-2-methyl-4-thiazolyl, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenecarboxylic acid #
5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-carboxylic acid

Specifications

400715-45-9
100.0
Amber glass bottle
MFCD02682053
AEDWMWQOFJYAGN-UHFFFAOYSA-N
5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-carboxylic acid
614467
97%
96.5
97%
C9H7NO2S2
5-2-methyl-1,3-thiazol-4-yl thiophene-2-carboxylic acid, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenecarboxylic acid, 2-thiophenecarboxylicacid, 5-2-methyl-4-thiazolyl, 5-2-methylthiazol-4-yl thiophene-2-carboxylic acid, 5-2-methyl-1,3-thiazol-4-yl thiophen-2-carboxylic acid, 5-2-methylthiazol-4-yl-thiophene-2-carboxylic acid, 5-2-methylthiazol4-yl-thiophene-2-carboxylic acid, thiophene-2-carboxylic acid, 5-2-methyl-4-thiazolyl, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenecarboxylic acid #
CC1=NC(=CS1)C2=CC=C(S2)C(=O)O
225.28
225.29
5-(2-Methyl-1, 3-thiazol-4-yl)-2-thiophenecarboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents