missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Methyl-1,3-thiazol-4-yl)-2-thiophenecarboxylic acid, 97%, Thermo Scientific™

Catalog No. CC18201CB
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC18201CB 250mg
CC18201DA 1g
CC18201DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC18201CB Supplier Thermo Scientific Chemicals Supplier No. CC18201CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 400715-45-9
Molecular Formula C9H7NO2S2
Molecular Weight (g/mol) 225.28
MDL Number MFCD02682053
InChI Key AEDWMWQOFJYAGN-UHFFFAOYSA-N
Synonym 5-2-methyl-1,3-thiazol-4-yl thiophene-2-carboxylic acid, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenecarboxylic acid, 2-thiophenecarboxylicacid, 5-2-methyl-4-thiazolyl, 5-2-methylthiazol-4-yl thiophene-2-carboxylic acid, 5-2-methyl-1,3-thiazol-4-yl thiophen-2-carboxylic acid, 5-2-methylthiazol-4-yl-thiophene-2-carboxylic acid, 5-2-methylthiazol4-yl-thiophene-2-carboxylic acid, thiophene-2-carboxylic acid, 5-2-methyl-4-thiazolyl, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenecarboxylic acid #
PubChem CID 614467
IUPAC Name 5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-carboxylic acid
SMILES CC1=NC(=CS1)C2=CC=C(S2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250mg
Formula Weight 225.29
Percent Purity 97%
Chemical Name or Material 5-(2-Methyl-1, 3-thiazol-4-yl)-2-thiophenecarboxylic acid
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.