missing translation for 'onlineSavingsMsg'
Learn More

[5-(2-Methyl-1,3-thiazol-4-yl)-2-thienyl]methanol 95%, Thermo Scientific™

Catalog No. CC18209DA
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC18209DA 1 g
CC18209DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC18209DA Supplier Thermo Scientific Chemicals Supplier No. CC18209DA

Chemical Identifiers

CAS 337508-70-0
Molecular Formula C9H9NOS2
Molecular Weight (g/mol) 211.297
InChI Key CQCKNGVVCKWDLF-UHFFFAOYSA-N
Synonym 5-2-methyl-1,3-thiazol-4-yl-2-thienyl methanol, 5-2-methyl-1,3-thiazol-4-yl thiophen-2-yl methanol, 5-2-methyl-4-thiazolyl thiophene-2-methanol, 5-2-methylthiazol-4-yl thiophen-2-yl methanol, 5-2-methyl-1,3-thiazol-4-yl thien-2-yl methanol, 5-2-methyl-1,3-thiazol-4-yl-2-thienyl methan-1-ol
PubChem CID 2776470
IUPAC Name [5-(2-methyl-1,3-thiazol-4-yl)thiophen-2-yl]methanol
SMILES CC1=NC(=CS1)C2=CC=C(S2)CO

Specifications

Assay Percent Range 95%
Quantity 1 g
Formula Weight 211.31
Percent Purity 95%
Chemical Name or Material [5-(2-Methyl-1, 3-thiazol-4-yl)-2-thienyl]methanol