missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Furyl)pyridine-2-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC55101DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC55101DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

930110-99-9
189.17
LTWJYKGQIACFMN-UHFFFAOYSA-N
26343608
C1=COC(=C1)C2=CN=C(C=C2)C(=O)O
C10H7NO3
MFCD10700052
5-furan-2-yl picolinic acid, 5-furan-2-yl pyridine-2-carboxylic acid, 5-2-furyl pyridine-2-carboxylic acid, 5-fur-2-yl pyridine-2-carboxylic acid, 5-furan-2-yl picolinicacid, 2-pyridinecarboxylicacid,5-2-furanyl, 2-pyridinecarboxylic acid, 5-2-furanyl
5-(furan-2-yl)pyridine-2-carboxylic acid

Specifications

5-(2-Furyl)pyridine-2-carboxylic acid
96.5
97%
C10H7NO3
MFCD10700052
LTWJYKGQIACFMN-UHFFFAOYSA-N
5-(furan-2-yl)pyridine-2-carboxylic acid
26343608
97%
930110-99-9
100.0
Amber Glass Bottle
1g
5-furan-2-yl picolinic acid, 5-furan-2-yl pyridine-2-carboxylic acid, 5-2-furyl pyridine-2-carboxylic acid, 5-fur-2-yl pyridine-2-carboxylic acid, 5-furan-2-yl picolinicacid, 2-pyridinecarboxylicacid,5-2-furanyl, 2-pyridinecarboxylic acid, 5-2-furanyl
C1=COC(=C1)C2=CN=C(C=C2)C(=O)O
189.17
189.17
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents