missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[5-(2-Furyl)-1H-pyrazol-3-yl]methanol, 97%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 5g |
| Formula Weight | 164.16 |
| Percent Purity | 97% |
| Chemical Name or Material | [5-(2-Furyl)-1H-pyrazol-3-yl]methanol |