missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]-7-pyrimidinone 98.0+%, TCI America™

Catalog No. p-7129351
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11345G 5 g
E113425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11345G Supplier TCI America Supplier No. E11345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 139756-21-1
Molecular Formula C17H20N4O2
Molecular Weight (g/mol) 312.37
MDL Number MFCD03093929
InChI Key MXQUEDUMKWBYHI-UHFFFAOYSA-N
PubChem CID 899745
IUPAC Name 5-(2-ethoxyphenyl)-1-methyl-3-propyl-1H,4H,7H-pyrazolo[4,3-d]pyrimidin-7-one
SMILES CCCC1=NN(C)C2=C1NC(=NC2=O)C1=CC=CC=C1OCC

Specifications

Melting Point 145°C
Quantity 5 g
Formula Weight 312.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(2-Ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]-7-pyrimidinone