missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Bromophenyl)-1,3-oxazole, ≥97%, Thermo Scientific™

$163.62 - $163.62

Chemical Identifiers

CAS 328270-70-8
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.057
MDL Number MFCD05668985
InChI Key JLTHLCLAPCIKJJ-UHFFFAOYSA-N
Synonym 5-2-bromophenyl oxazole, 5-2-bromophenyl-1,3-oxazole, oxazole,5-2-bromophenyl
PubChem CID 3780551
IUPAC Name 5-(2-bromophenyl)-1,3-oxazole
SMILES C1=CC=C(C(=C1)C2=CN=CO2)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC35710DA
SDS View Documents
Thermo Scientific Chemicals
CC35710DA
1 g
Each for $163.62
Only null left
 
Description

Description

Specifications

Chemical Identifiers

328270-70-8
224.057
JLTHLCLAPCIKJJ-UHFFFAOYSA-N
3780551
C1=CC=C(C(=C1)C2=CN=CO2)Br
C9H6BrNO
MFCD05668985
5-2-bromophenyl oxazole, 5-2-bromophenyl-1,3-oxazole, oxazole,5-2-bromophenyl
5-(2-bromophenyl)-1,3-oxazole

Specifications

328270-70-8
100.0
Amber glass bottle
MFCD05668985
5-2-bromophenyl oxazole, 5-2-bromophenyl-1,3-oxazole, oxazole,5-2-bromophenyl
C1=CC=C(C(=C1)C2=CN=CO2)Br
224.057
224.06
5-(2-Bromophenyl)-1, 3-oxazole
94.5
95%
C9H6BrNO
1 g
JLTHLCLAPCIKJJ-UHFFFAOYSA-N
5-(2-bromophenyl)-1,3-oxazole
3780551
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'