missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Bromophenyl)-1,3-oxazole, ≥97%, Thermo Scientific™

Catalog No. CC35710DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC35710DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC35710DA Supplier Thermo Scientific Chemicals Supplier No. CC35710DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 328270-70-8
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.057
MDL Number MFCD05668985
InChI Key JLTHLCLAPCIKJJ-UHFFFAOYSA-N
Synonym 5-2-bromophenyl oxazole, 5-2-bromophenyl-1,3-oxazole, oxazole,5-2-bromophenyl
PubChem CID 3780551
IUPAC Name 5-(2-bromophenyl)-1,3-oxazole
SMILES C1=CC=C(C(=C1)C2=CN=CO2)Br

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 224.06
Percent Purity 95%
Chemical Name or Material 5-(2-Bromophenyl)-1, 3-oxazole
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.