missing translation for 'onlineSavingsMsg'
Learn More

5-(2,5-Dioxotetrahydrofuryl)-3-methyl-3-cyclohexene-1,2-dicarboxylic Anhydride 95.0+%, TCI America™

$136.08 - $1,163.17

Chemical Identifiers

CAS 73003-90-4
Molecular Formula C13H12O6
Molecular Weight (g/mol) 264.23
MDL Number MFCD00039141
InChI Key DGQOZCNCJKEVOA-UHFFFAOYNA-N
PubChem CID 175380
IUPAC Name 5-(2,5-dioxooxolan-3-yl)-7-methyl-1,3,3a,4,5,7a-hexahydro-2-benzofuran-1,3-dione
SMILES CC1=CC(CC2C1C(=O)OC2=O)C1CC(=O)OC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D190125G
View Documents
TCI America
D190125G
25 g N/A
Only null left
 
D1901500G
View Documents
TCI America
D1901500G
500 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

73003-90-4
264.23
DGQOZCNCJKEVOA-UHFFFAOYNA-N
5-(2,5-dioxooxolan-3-yl)-7-methyl-1,3,3a,4,5,7a-hexahydro-2-benzofuran-1,3-dione
C13H12O6
MFCD00039141
175380
CC1=CC(CC2C1C(=O)OC2=O)C1CC(=O)OC1=O

Specifications

73003-90-4
White-Yellow
MFCD00039141
DGQOZCNCJKEVOA-UHFFFAOYNA-N
5-(2,5-dioxooxolan-3-yl)-7-methyl-1,3,3a,4,5,7a-hexahydro-2-benzofuran-1,3-dione
175380
≥95.0% (T)
5-(2,5-Dioxotetrahydrofuryl)-3-methyl-3-cyclohexene-1,2-dicarboxylic Anhydride
174°C
C13H12O6
25 g
CC1=CC(CC2C1C(=O)OC2=O)C1CC(=O)OC1=O
264.23
264.23
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-5435

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'