missing translation for 'onlineSavingsMsg'
Learn More

5-(2,3-Dichlorophenyl)-1H-tetrazole, 97%, Thermo Scientific Chemicals

Catalog No. p-7025449
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2092106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2092106 Supplier Thermo Scientific Chemicals Supplier No. B2092106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Soluble in water 0.026 g/L (25°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 175205-12-6
Molecular Formula C7H3Cl2N4
Molecular Weight (g/mol) 214.03
MDL Number MFCD00068096,MFCD04035663
InChI Key IQJBSNPEVBTDMM-UHFFFAOYSA-N
Synonym 5-2,3-dichlorophenyl-1h-tetrazole, 5-2,3-dichlorophenyl tetrazole, 5-2,3-dichlorophenyl-2h-tetrazole, 5-2,3-dichlorophenyl-1h-1,2,3,4-tetrazole, 2h-tetrazole,5-2,3-dichlorophenyl, 5-2,3-dichlorophenyl-1h-1,2,3,4-tetraazole, maybridge1_006937, acmc-20ama8, 5-2,3-dichlorophenyl-1h-tetraazole, 5-2,3-dichloro-phenyl-1h-tetrazole
PubChem CID 4690559
IUPAC Name 5-(2,3-dichlorophenyl)-2H-tetrazole
SMILES ClC1=CC=CC(C2=N[N-]N=N2)=C1Cl

Specifications

Melting Point 180°C to 182°C
Quantity 5 g
Solubility Information Soluble in water 0.026g/L (25°C).
Formula Weight 215.04
Percent Purity 97%
Chemical Name or Material 5-(2,3-Dichlorophenyl)-1H-tetrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only