missing translation for 'onlineSavingsMsg'
Learn More

5,12-Naphthacenequinone 98.0+%, TCI America™

Catalog No. p-7133889
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N060310G 10 g
N060325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N060310G Supplier TCI America Supplier No. N060310G

Chemical Identifiers

CAS 1090-13-7
Molecular Formula C18H10O2
Molecular Weight (g/mol) 258.28
MDL Number MFCD00003701
InChI Key LZPBKINTWROMEA-UHFFFAOYSA-N
Synonym 5,12-naphthacenequinone, 5,12-naphthacenedione, naphthacenequinone, 5,12-tetracenequinone, tetracenemonoquinone, naphthacene-6,11-quinone, naphthacene-5,12-dione, tetracenequinone, 5,12-naphtacenequinone, tetracene-5,12-quinone
PubChem CID 14160
ChEBI CHEBI:51287
IUPAC Name 5,12-dihydrotetracene-5,12-dione
SMILES O=C1C2=CC=CC=C2C(=O)C2=C1C=C1C=CC=CC1=C2

Specifications

Quantity 10 g
Formula Weight 258.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5,12-Naphthacenequinone