missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,10-Dihydro-5,10-dimethylphenazine 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 153°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 210.28 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5,10-Dihydro-5,10-dimethylphenazine |
Safety and Handling
| TSCA | No |