missing translation for 'onlineSavingsMsg'
Learn More

5,10,15,20-Tetrakis(pentafluorophenyl)-21H,23H-porphine, 95%

Catalog No. AC292151000
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC292151000 100 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC292151000 Supplier Thermo Scientific Chemicals Supplier No. 292151000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25440-14-6 | C44H10F20N4 | 974.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25440-14-6
Molecular Formula C44H10F20N4
Molecular Weight (g/mol) 974.56
MDL Number MFCD00010032
InChI Key GBAPBSUXEFFUAD-UHFFFAOYSA-N
Synonym 5,10,15,20-tetrakis pentafluorophenyl porphyrin, 5,10,15,20-tetrakis pentafluorophenyl-21h,23h-porphine, meso-tetra pentafluorophenyl porphine, 21h,23h-porphine, 5,10,15,20-tetrakis pentafluorophenyl, 5,10,15,20-tetrakis 2,3,4,5,6-pentafluorophenyl-21,22-dihydroporphyrin, 2,7,12,17-tetrakis 2,3,4,5,6-pentafluorophenyl-21,22,23,24-tetraazapentacyclo 16.2.1.1 3 ,?.1?, 1 1 .1 1 3 , 1 ? tetracosa-1,3,5,7,9,11 23 ,12,14,16,18 21 ,19-undecaene, acmc-209gjw, iron iii ionophore iv, tetra pentafluorophenyl poephine, tetrakis pentafluorophenyl porphyrin
PubChem CID 3671775
IUPAC Name 5,10,15,20-tetrakis(2,3,4,5,6-pentafluorophenyl)-21,22-dihydroporphyrin
SMILES FC1=C(F)C(F)=C(C(F)=C1F)C1=C2NC(C=C2)=C(C2=NC(C=C2)=C(C2=NC(C=C2)=C(C2=CC=C1N2)C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Purple or Violet
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Formula Weight 974.57
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 5,10,15,20-Tetrakis(pentafluorophenyl)-21H,23H-porphine
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only