missing translation for 'onlineSavingsMsg'
Learn More

5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin 97.0+%, TCI America™

Catalog No. T1360100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1360100MG 100 mg
T13601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1360100MG Supplier TCI America Supplier No. T1360100MG
Only null left

Chemical Identifiers

CAS 22112-78-3
Molecular Formula C48H38N4O4
Molecular Weight (g/mol) 734.856
MDL Number MFCD00012072
InChI Key SKXWDGDNWBYACJ-UHFFFAOYSA-N
Synonym 5,10,15,20-tetrakis 4-methoxyphenyl-21h,23h-porphine, 5,10,15,20-tetrakis 4-methoxyphenyl porphyrin, tetra p-methoxyphenyl porphyrin, 21h,23h-porphine, 5,10,15,20-tetrakis 4-methoxyphenyl, 5,10,15,20-tetrakis 4-methoxyphenyl-21,22-dihydroporphyrin, 2,7,12,17-tetrakis 4-methoxyphenyl-21,22,23,24-tetraazapentacyclo 16.2.1.1 3 ,?.1?, 1 1 .1 1 3 , 1 ? tetracosa-1,3,5,7,9,11 23 ,12,14,16,18 21 ,19-undecaene, 4-methoxy-1-7,12,17-tris 4-methoxyphenyl-21,22,23,24-tetraazapentacyclo 16.2 .1.1<3,6>.1<8,11>.1<13,16> tetracosa-1,3,5,7,9,11 23 ,12,14,16,18 21 ,19-undec aen-2-yl benzene, acmc-209fs6, tetrakis p-methoxyphenyl porphyrin, meso-tetra 4-methoxyphenyl porphine
PubChem CID 140886
IUPAC Name 5,10,15,20-tetrakis(4-methoxyphenyl)-21,22-dihydroporphyrin
SMILES COC1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=C(C=C7)OC)C8=CC=C(C=C8)OC)C9=CC=C(C=C9)OC)N3

Specifications

Color Blue-Purple
Quantity 100 mg
Formula Weight 734.86
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin