missing translation for 'onlineSavingsMsg'
Learn More

5,10,15,20-Tetrakis(3,5-dihydroxyphenyl)porphyrin 90.0+%, TCI America™

Catalog No. p-7136975
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1815100MG 100 mg
T18151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1815100MG Supplier TCI America Supplier No. T1815100MG

Chemical Identifiers

CAS 145764-54-1
Molecular Formula C44H30N4O8
Molecular Weight (g/mol) 742.744
MDL Number MFCD02093497
InChI Key XDVQXKKXBXLGCO-UHFFFAOYSA-N
Synonym 5,10,15,20-Tetrakis(3,5-dihydroxyphenyl)-21H,23H-porphine
PubChem CID 471968
IUPAC Name 5-[10,15,20-tris(3,5-dihydroxyphenyl)-21,24-dihydroporphyrin-5-yl]benzene-1,3-diol
SMILES C1=CC2=C(C3=CC=C(N3)C(=C4C=CC(=N4)C(=C5C=CC(=N5)C(=C1N2)C6=CC(=CC(=C6)O)O)C7=CC(=CC(=C7)O)O)C8=CC(=CC(=C8)O)O)C9=CC(=CC(=C9)O)O

Specifications

Color Blue-Purple
Quantity 100 mg
Formula Weight 742.74
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5,10,15,20-Tetrakis(3,5-dihydroxyphenyl)porphyrin