missing translation for 'onlineSavingsMsg'
Learn More

5,10,15,20-Tetrakis(2,4,6-trimethylphenyl)porphyrin 98.0+%, TCI America™

Catalog No. T1729100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1729100MG 100 mg
T17291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1729100MG Supplier TCI America Supplier No. T1729100MG

Chemical Identifiers

CAS 56396-12-4
Molecular Formula C56H54N4
Molecular Weight (g/mol) 783.076
MDL Number MFCD01321200
InChI Key NVRVXKGQRXOICG-UHFFFAOYSA-N
Synonym 5,10,15,20-Tetrakis(2,4,6-trimethylphenyl)-21H,23H-porphine
PubChem CID 481541
IUPAC Name 5,10,15,20-tetrakis(2,4,6-trimethylphenyl)-21,22-dihydroporphyrin
SMILES CC1=CC(=C(C(=C1)C)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=C(C=C(C=C7C)C)C)C8=C(C=C(C=C8C)C)C)C9=C(C=C(C=C9C)C)C)N3)C

Specifications

Color Purple
Quantity 100 mg
Formula Weight 783.08
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5,10,15,20-Tetrakis(2,4,6-trimethylphenyl)porphyrin