missing translation for 'onlineSavingsMsg'
Learn More

5-(1-Piperidinylmethyl)thiophene-2-boronic acid pinacol ester, Thermo Scientific Chemicals

$275.38 - $275.38

Chemical Identifiers

CAS 1218790-44-3
Molecular Formula C16H26BNO2S
Molecular Weight (g/mol) 307.259
MDL Number MFCD11113036
InChI Key KDPVLCCGXRCQCV-UHFFFAOYSA-N
Synonym 5-1-piperidinylmethyl thiophene-2-boronic acid pinacol ester, 1-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl piperidine, 5-piperidine methyl-2-thiopheneboronic acid pinacol ester, 1-5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl piperidine, amtb125, 5-piperidinomethyl-2-thiopheneboronic acid pinacol ester, 5-1-piperidinylmethyl-thiophene-2-boronic acid pinacol ester, 1-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-thiophen-2-ylmethyl-piperidine
PubChem CID 46739745
IUPAC Name 1-[[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl]piperidine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(S2)CN3CCCCC3
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5111706
View Documents
Thermo Scientific Chemicals
H5111706
5 g
Each for $275.38
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1218790-44-3
307.259
KDPVLCCGXRCQCV-UHFFFAOYSA-N
46739745
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(S2)CN3CCCCC3
C16H26BNO2S
MFCD11113036
5-1-piperidinylmethyl thiophene-2-boronic acid pinacol ester, 1-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl piperidine, 5-piperidine methyl-2-thiopheneboronic acid pinacol ester, 1-5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl piperidine, amtb125, 5-piperidinomethyl-2-thiopheneboronic acid pinacol ester, 5-1-piperidinylmethyl-thiophene-2-boronic acid pinacol ester, 1-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-thiophen-2-ylmethyl-piperidine
1-[[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl]piperidine

Specifications

1218790-44-3
MFCD11113036
5-1-piperidinylmethyl thiophene-2-boronic acid pinacol ester, 1-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl piperidine, 5-piperidine methyl-2-thiopheneboronic acid pinacol ester, 1-5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl piperidine, amtb125, 5-piperidinomethyl-2-thiopheneboronic acid pinacol ester, 5-1-piperidinylmethyl-thiophene-2-boronic acid pinacol ester, 1-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-thiophen-2-ylmethyl-piperidine
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(S2)CN3CCCCC3
307.259
307.26
C16H26BNO2S
5 g
KDPVLCCGXRCQCV-UHFFFAOYSA-N
1-[[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl]piperidine
46739745
5-(1-Piperidinylmethyl)thiophene-2-boronic acid pinacol ester
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only