missing translation for 'onlineSavingsMsg'
Learn More

5-(1-Piperidinylmethyl)thiophene-2-boronic acid pinacol ester, Thermo Scientific Chemicals

Catalog No. AAH5111706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5111706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5111706 Supplier Thermo Scientific Chemicals Supplier No. H5111706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1218790-44-3
Molecular Formula C16H26BNO2S
Molecular Weight (g/mol) 307.259
MDL Number MFCD11113036
InChI Key KDPVLCCGXRCQCV-UHFFFAOYSA-N
Synonym 5-1-piperidinylmethyl thiophene-2-boronic acid pinacol ester, 1-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl piperidine, 5-piperidine methyl-2-thiopheneboronic acid pinacol ester, 1-5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl piperidine, amtb125, 5-piperidinomethyl-2-thiopheneboronic acid pinacol ester, 5-1-piperidinylmethyl-thiophene-2-boronic acid pinacol ester, 1-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-thiophen-2-ylmethyl-piperidine
PubChem CID 46739745
IUPAC Name 1-[[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl]piperidine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(S2)CN3CCCCC3

Specifications

Quantity 5 g
Formula Weight 307.26
Chemical Name or Material 5-(1-Piperidinylmethyl)thiophene-2-boronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only