missing translation for 'onlineSavingsMsg'
Learn More

5-(1,3-Dioxolan-2-yl)-2-furaldehyde 95.0+%, TCI America™

Catalog No. D24181G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D24181G Supplier TCI America Supplier No. D24181G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 117953-13-6
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00671521
InChI Key XTPZCGOSUHCSQZ-UHFFFAOYSA-N
Synonym 5-(1,3-Dioxolan-2-yl)furfural
PubChem CID 14616597
IUPAC Name 5-(1,3-dioxolan-2-yl)furan-2-carbaldehyde
SMILES O=CC1=CC=C(O1)C1OCCO1

Specifications

Melting Point 41°C
Quantity 1 g
Formula Weight 168.15
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-(1,3-Dioxolan-2-yl)-2-furaldehyde
TSCA No
Recommended Storage Refrigerator