missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone, 99%
CAS: 77943-39-6 | C10H11NO2 | 177.2 g/mol
$95.02 - $95.02
Chemical Identifiers
| CAS | 77943-39-6 |
|---|---|
| Molecular Formula | C10H11NO2 |
| Molecular Weight (g/mol) | 177.2 |
| MDL Number | MFCD00010845 |
| InChI Key | PPIBJOQGAJBQDF-VXNVDRBHSA-N |
| Synonym | 4r,5s-+-4-methyl-5-phenyl-2-oxazolidinone, 4r,5s-4-methyl-5-phenyloxazolidin-2-one, 4r,5s-4-methyl-5-phenyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-methyl-5-phenyl-, 4r,5s, 4r,5s-4-methyl-5-phenyl-2-oxazolidinone, meph-2-oxazolidinone, pubchem6871, 2-oxazolidinone, 4-methyl-5-phenyl-, 4r-cis, 4-methyl-5-phenyl-2-oxazolidinone 4r-cis, 4alpha-methyl-5alpha-phenyl-2-oxazolidinone |
| PubChem CID | 2733812 |
| IUPAC Name | (4R,5S)-4-methyl-5-phenyl-1,3-oxazolidin-2-one |
| SMILES | CC1C(OC(=O)N1)C2=CC=CC=C2 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
|
AC337630010
|
Thermo Scientific Chemicals
337630010 |
1 g | Glass Bottle |
Each for $95.02
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 77943-39-6 | |
| 177.2 | |
| PPIBJOQGAJBQDF-VXNVDRBHSA-N | |
| 2733812 | |
| CC1C(OC(=O)N1)C2=CC=CC=C2 |
| C10H11NO2 | |
| MFCD00010845 | |
| 4r,5s-+-4-methyl-5-phenyl-2-oxazolidinone, 4r,5s-4-methyl-5-phenyloxazolidin-2-one, 4r,5s-4-methyl-5-phenyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-methyl-5-phenyl-, 4r,5s, 4r,5s-4-methyl-5-phenyl-2-oxazolidinone, meph-2-oxazolidinone, pubchem6871, 2-oxazolidinone, 4-methyl-5-phenyl-, 4r-cis, 4-methyl-5-phenyl-2-oxazolidinone 4r-cis, 4alpha-methyl-5alpha-phenyl-2-oxazolidinone | |
| (4R,5S)-4-methyl-5-phenyl-1,3-oxazolidin-2-one |
Specifications
| 118°C to 121°C | |
| 98.5 | |
| Brown-Yellow to Colorless | |
| 98.5% min. (HPLC) | |
| Glass Bottle | |
| + 162 | |
| PPIBJOQGAJBQDF-VXNVDRBHSA-N | |
| (4R,5S)-4-methyl-5-phenyl-1,3-oxazolidin-2-one | |
| 1 g | |
| 2733812 | |
| 99% | |
| (4R, 5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone |
| 77943-39-6 | |
| 100.0 | |
| Authentic | |
| C10H11NO2 | |
| MFCD00010845 | |
| 4r,5s-+-4-methyl-5-phenyl-2-oxazolidinone, 4r,5s-4-methyl-5-phenyloxazolidin-2-one, 4r,5s-4-methyl-5-phenyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-methyl-5-phenyl-, 4r,5s, 4r,5s-4-methyl-5-phenyl-2-oxazolidinone, meph-2-oxazolidinone, pubchem6871, 2-oxazolidinone, 4-methyl-5-phenyl-, 4r-cis, 4-methyl-5-phenyl-2-oxazolidinone 4r-cis, 4alpha-methyl-5alpha-phenyl-2-oxazolidinone | |
| CC1C(OC(=O)N1)C2=CC=CC=C2 | |
| + 162.00 (20.00°C c=2,CHCL3) | |
| 177.2 | |
| 177.2 | |
| Liquid |
RUO – Research Use Only