Chemglass Inc Reaction Vessel Lid, 4-Neck, 24/40 Center Neck, 3-24/40 Side Necks, 100mm Schott Flange
Catalog No. CG194101
Reaction Vessel Lid, 4-Neck, 24/40 Center Neck, 3-24/40 Side Necks, 100mm Schott Flange. Four neck vessel lid to fit CG-1920 through CG-1936 vessels. Lid has a flat ground surface allowing it to be greased or used with an o-ring. Note: Four neck lids are available with joint sizes and configurations other than those listed. Please call for details.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Reaction Vessel Lid, 4-Neck, 24/40 Center Neck, 3-24/40 Side Necks, 100mm Schott Flange. Four neck vessel lid to fit CG-1920 through CG-1936 vessels. Lid has a flat ground surface allowing it to be greased or used with an o-ring. Note: Four neck lids are available with joint sizes and configurations other than those listed. Please call for details.