missing translation for 'onlineSavingsMsg'
Learn More

4N6FLOQSwabs™, regular tip, peelpouch

Catalog No. 501607765
Encompass
Change view
Click to view available options
Quantity:
100 swabs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501607765 100 swabs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501607765 Supplier Thermo Fisher Scientific Supplier No. 4473979

4N6FLOQSwabs, Regular Tip are comprised of a solid, molded plastic applicator shaft with a regular-sized tip that can vary in size and shape. The tip of the applicator is coated with short Nylon fibers that are arranged in a perpendicular fashion. This perpendicular arrangement results from a process called flocking, where the fibers are sprayed onto the tip of the swab while it is held in an electrostatic field. This process creates a highly absorbent, thin layer with an open structure.

Key Features:
• Certified free of DNase, RNase and amplifiable human DNA
• Rapid absorption capability and superior sample release
• Regular size tip for buccal cell collection
• Individually packaged in a convenient peel pouch format
• Break Point (from tip end): 20 mm

Unlike traditional fiber wound swabs, 4N6FLOQSwabs have no internal absorbent core to disperse and entrap the specimen—the sample stays close to the surface for faster and more efficient elution. The perpendicular Nylon fibers act like a soft brush that facilitates improved collection of cellular material. Capillary action between the fiber strands facilitates strong hydraulic uptake of liquid sample, and the sample stays close to the surface allowing easy elution.

4N6FLOQSwabs are manufactured by Copan, a market leader in the area of sample collection tools, and distributed globally by Thermo Fisher Scientific.

For Forensic or Paternity Use Only.

Specifications

Quantity 100 swabs
Content And Storage 100 FLOQSwabs Swabs
Store at room temperature
Product Line 4N6FLOQSwabs
Shipping Condition Room Temperature
Product Type Swab
Tip Material Nylon
Tip Type Regular