missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4N6FLOQSwabs™ for Crime Scene, regular tip, peelpouch
Specifications
Specifications
| Tip Type | Regular |
| Content And Storage | 100 Regular head tip FLOQSwabs™, in peelpouch, with Antimicrobial Action Store at RT |
| Applicator Material | Nylon |
| Product Line | 4N6FLOQSwabs |
| Shipping Condition | Approved for shipment at Room Temperature or on Dry Ice |
| Format | Pouch |
| Quantity | Case of 100 |