Learn More
Alkali Scientific 4mm Electroporation Cuvettes, Round Lid, Sterile, 50/cs
Catalog No. 502389365
Encompass
Manufactured from high quality durable polycarbonate material which has been tested to optimize transfection efficiencies. Precision built to give exacting tolerances ensuring reproducible results. Polished aluminum plates reduce static and arcing for optimum performance. Individually wrapped and gamma irradiated providing guaranteed sterility. Compatible with most Electroporation devices. Individually color coded cuvette caps identify gap widths of 4mm. Each bag contains 50 individually wrapped sterile cuvettes.
Catalog No. 502389365 Supplier Alkali Scientific Supplier No. EP104004
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Manufactured from high quality durable polycarbonate material which has been tested to optimize transfection efficiencies. Precision built to give exacting tolerances ensuring reproducible results. Polished aluminum plates reduce static and arcing for optimum performance. Individually wrapped and gamma irradiated providing guaranteed sterility. Compatible with most Electroporation devices. Individually color coded cuvette caps identify gap widths of 4mm. Each bag contains 50 individually wrapped sterile cuvettes.