missing translation for 'onlineSavingsMsg'
Learn More

4H-Thieno[3,2-b]pyrrole-5-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog No. AAH5820003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5820003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5820003 Supplier Thermo Scientific Chemicals Supplier No. H5820003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 39793-31-2 | Molecular Formula: C7H5NO2S | Molecular Weight: 167.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39793-31-2
Molecular Formula C7H5NO2S
Molecular Weight (g/mol) 167.182
MDL Number MFCD07368542
InChI Key PMHDSACGRKBACK-UHFFFAOYSA-N
Synonym 4h-thieno 3,2-b pyrrole-5-carboxylic acid, 4h-thieno 3,2-b pyrole-5-carboxylic acid, 4wl, d-amino acid oxidase inhibitor, 4h-thieno 3,2-b pyrrole-5-carboxylicacid, thiopheno 3,2-b pyrrole-5-carboxylic acid, 4h-thieno 2,3-d pyrrole-5-carboxylic acid, 4h-thieno 3 pound not2-b pyrrole-5-carboxylic acid
PubChem CID 4961254
IUPAC Name 4H-thieno[3,2-b]pyrrole-5-carboxylic acid
SMILES C1=CSC2=C1NC(=C2)C(=O)O

Specifications

Melting Point 180°C
Quantity 1 g
Formula Weight 167.19
Percent Purity 97%
Chemical Name or Material 4H-Thieno[3,2-b]pyrrole-5-carboxylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only