missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4H-Pyran-4-one 98.0+%, TCI America™
$342.98 - $1371.95
Chemical Identifiers
| CAS | 108-97-4 |
|---|---|
| Molecular Formula | C5H4O2 |
| Molecular Weight (g/mol) | 96.085 |
| MDL Number | MFCD00006576 |
| InChI Key | CVQUWLDCFXOXEN-UHFFFAOYSA-N |
| Synonym | gamma-Pyrone |
| PubChem CID | 7968 |
| ChEBI | CHEBI:37966 |
| IUPAC Name | pyran-4-one |
| SMILES | C1=COC=CC1=O |
Chemical Identifiers
| 108-97-4 | |
| 96.085 | |
| CVQUWLDCFXOXEN-UHFFFAOYSA-N | |
| 7968 | |
| pyran-4-one |
| C5H4O2 | |
| MFCD00006576 | |
| gamma-Pyrone | |
| CHEBI:37966 | |
| C1=COC=CC1=O |
Specifications
| 108-97-4 | |
| White-Yellow | |
| C5H4O2 | |
| 1 g | |
| CVQUWLDCFXOXEN-UHFFFAOYSA-N | |
| pyran-4-one | |
| 7968 | |
| 96.09 | |
| Crystalline Lumps |
| 32°C | |
| 215°C | |
| MFCD00006576 | |
| gamma-Pyrone | |
| C1=COC=CC1=O | |
| 96.085 | |
| CHEBI:37966 | |
| ≥98.0% (GC) | |
| 4H-Pyran-4-one |
Safety and Handling
TSCA : No