missing translation for 'onlineSavingsMsg'
Learn More

4H-Pyran-4-one 98.0+%, TCI America™

Catalog No. P14505G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14505G 5 g
P14501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14505G Supplier TCI America Supplier No. P14505G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-97-4
Molecular Formula C5H4O2
Molecular Weight (g/mol) 96.085
MDL Number MFCD00006576
InChI Key CVQUWLDCFXOXEN-UHFFFAOYSA-N
Synonym gamma-Pyrone
PubChem CID 7968
ChEBI CHEBI:37966
IUPAC Name pyran-4-one
SMILES C1=COC=CC1=O

Specifications

Melting Point 32°C
Color White-Yellow
Boiling Point 215°C
Quantity 5 g
Formula Weight 96.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4H-Pyran-4-one