missing translation for 'onlineSavingsMsg'
Learn More

(4aS,8aR)-2-Benzoyloctahydro-6(2H)-isoquinolinone 90.0+%, TCI America™

Catalog No. B242050MG
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B242050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B242050MG Supplier TCI America Supplier No. B242050MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 52390-26-8
Molecular Formula C16H19NO2
Molecular Weight (g/mol) 257.33
MDL Number MFCD08276277
InChI Key OSZFDJVZOKTWRW-UHFFFAOYNA-N
PubChem CID 12262835
IUPAC Name 2-benzoyl-decahydroisoquinolin-6-one
SMILES O=C(N1CCC2CC(=O)CCC2C1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 50 mg
Formula Weight 257.33
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (4aS,8aR)-2-Benzoyloctahydro-6(2H)-isoquinolinone