missing translation for 'onlineSavingsMsg'
Learn More

KleenGuard™ G35 Seamless Knit Nylon Inspection Gloves

Catalog No. 19167728
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19167728 Medium
19167729 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19167728 Supplier KleenGuard™ Supplier No. 38718

Low-lint gloves are ideal for detecting imperfections and defects when conducting critical quality control inspections. Ambidextrous KleenGuard™ G35 Seamless Knit Nylon Inspection Gloves are silicone-free for use in a wide variety of applications including film handling, aerospace, and automotive assembly.

  • Ambidextrous design allows gloves to be replaced one at a time, rather than in pairs
  • High degree of sensitivity and dexterity for inspecting for defects and imperfections
  • Designed for critical quality control inspections
  • Seamless knit nylon shell reduces skin irritation
  • Low lint for cleanroom applications
  • Silicone-free

Specifications

Construction Seamless Knit
Coating Material Uncoated
Color White
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Nylon
Product Type Inspection Gloves
Size Medium
Sterility Non-sterile
For Use With (Application) Automotive Assembly, Aerospace Engineering, and Quality Control Inspections
Glove Cuff Style Knit
Series KleenGuard™ G35
Show More Show Less