missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boekel Scientific™ Open Source Bio™ (OSB) Transmitter: Probes
For use with Open Source Bio (OSB) Transmitter.
$40.24 - $704.39
| Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
|
11700160
|
Boekel Scientific™
9510001 |
pH Probe |
Each for $237.50
|
|
|||||
|
11700162
|
Boekel Scientific™
9510003 |
DO Probe |
Each for $704.39
|
|
|||||
|
11700161
|
Boekel Scientific™
BKL9510002 |
Conductivity Probe (type k=1.0) |
Each for $322.00
|
|
|||||
|
11700163
|
Boekel Scientific™
9510004 |
Temperature Probe |
Each for $40.24
|
|
|||||