Learn More
Justrite™ Explosion-Proof Air Conditioners
Accessory for Outdoor Safety Lockers
$36,648.04
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
|
19414037
|
Justrite™
915307 |
12-,16-drum fire rated Agri-Turf lockers |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- For use with 6-,9-,12-,16-drum fire-rated Agri-Turf™ lockers only
- Available in 4.4kWh (15000 BTU) and 5.9kWh (20000 BTU) energy capacity
Applications:
Adds air conditioning where comfort, chemical stability or freeze protection is important
†For optimum temperature control, appropriate size R-11 insulation package must be ordered when unit is purchased in conjunction with fire-rated models. This accessory must be purchased in conjunction with locker to ensure proper installation and to retain FM-approval.
Specifications
| Explosion-proof Air conditioner |