missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Biohazard Identification Signs: Biohazard Area

Catalog No. 19140988
Change view
Click to view available options
Material:
Aluminum
Plastic
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19140988 Plastic
19140994 Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19140988 Supplier Brady™ Supplier No. 126652

For identification of equipment, rooms and materials that contain, or are contaminated with, hazardous agents

  • Made from aluminum, plastic or self-sticking polyester
  • Size: 10 in. H x 14 in. W
  • Legend: BIOHAZARD AREA

Specifications

Material Plastic
Height (English) 10 in.
Height (Metric) 3 m
Width (English) 14 in.
Width (Metric) 4.2 m
Displayed Text BIOHAZARD AREA
Product Type Biohazard Identification Sign